siRNA Details
| Gene Symbol: STXBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178509.5 | NP_848604.3 | 4 | 342 | ||
| XM_005257187.4 | XP_005257244.1 | 4 | 378 | |||
| XM_006721797.3 | XP_006721860.1 | 4 | 376 | |||
| XM_006721798.3 | XP_006721861.1 | 4 | 373 | |||
| XM_017024410.1 | XP_016879899.1 | 4 | 379 | |||
| XM_017024411.1 | XP_016879900.1 | 4 | 377 | |||
| XM_017024412.1 | XP_016879901.1 | 4 | 376 | |||
| XM_017024414.1 | XP_016879903.1 | 4 | 378 | |||
| XM_017024415.1 | XP_016879904.1 | 4 | 378 | |||
| GenBank mRNA | AK122865.1 | - | 4 | 342 | ||
| BC125116.1 | - | 4 | 331 | 40118892 | ||

STXBP4
Genomic Map