siRNA Details
| Gene Symbol: HCG27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026791.1 | 2 | 1521 | |||
| GenBank mRNA | AK097442.1 | - | 2 | 1521 | ||
| BC041465.2 | - | 1 | 1400 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCG27| Gene Symbol: HCG27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026791.1 | 2 | 1521 | |||
| GenBank mRNA | AK097442.1 | - | 2 | 1521 | ||
| BC041465.2 | - | 1 | 1400 | |||