siRNA Details
| Gene Symbol: EPHX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173567.4 | NP_775838.3 | 7 | 1092 | ||
| GenBank mRNA | AK074822.1 | - | 7 | 1052 | ||
| BC041475.1 | - | 7 | 1067 | 5244314 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPHX4| Gene Symbol: EPHX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173567.4 | NP_775838.3 | 7 | 1092 | ||
| GenBank mRNA | AK074822.1 | - | 7 | 1052 | ||
| BC041475.1 | - | 7 | 1067 | 5244314 | ||