siRNA Details
| Gene Symbol: IPMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152230.4 | NP_689416.1 | 5 | 919 | ||
| XM_011539565.2 | XP_011537867.1 | 5 | 960 | |||
| XM_017016013.1 | XP_016871502.1 | 5 | 586 | |||
| GenBank mRNA | AF432853.1 | - | 5 | 596 | ||
| AK096839.1 | - | 2 | 226 | |||
| BC065709.1 | - | 5 | 923 | 6654071 | ||

IPMK
Genomic Map