siRNA Details
| Gene Symbol: CXorf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169574.1 | NP_001163045.1 | 2, 3 | 452 | ||
| NM_152761.2 | NP_689974.2 | 2, 3 | 452 | |||
| XM_011545473.2 | XP_011543775.1 | 2, 3 | 209 | |||
| GenBank mRNA | AK058173.1 | - | 2, 3 | 452 | ||
| BC117344.1 | - | 2, 3 | 364 | 40125895 | ||
| BC117346.1 | - | 2, 3 | 364 | 40125897 | ||
| BC144007.1 | - | 2, 3 | 364 | 9052526 | ||

CXorf58
Genomic Map