siRNA Details
| Gene Symbol: FBXO33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203301.3 | NP_976046.1 | 3 | 1317 | ||
| GenBank mRNA | AK294140.1 | - | 1 | 161 | ||
| BC053537.1 | - | 3 | 1233 | 5181903 | ||
| BC068566.1 | - | 3 | 1289 | 5270962 | ||
| BX248772.1 | - | 3 | 971 | |||

FBXO33
Genomic Map