siRNA Details
| Gene Symbol: TTLL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130045.1 | NP_001123517.1 | 12 | 1305 | ||
| NM_153254.2 | NP_694986.2 | 8 | 1112 | |||
| XM_005244738.1 | XP_005244795.1 | 8 | 1139 | |||
| XM_011541177.2 | XP_011539479.1 | 12 | 1382 | |||
| XM_017000906.1 | XP_016856395.1 | 8 | 1383 | |||
| XM_017000907.1 | XP_016856396.1 | 9 | 1464 | |||
| XM_017000908.1 | XP_016856397.1 | 9 | 1464 | |||
| XM_017000909.1 | XP_016856398.1 | 9 | 1464 | |||
| XM_017000910.1 | XP_016856399.1 | 9 | 1464 | |||
| XM_017000911.1 | XP_016856400.1 | 9 | 1464 | |||
| XM_017000912.1 | XP_016856401.1 | 10 | 1236 | |||
| GenBank mRNA | AK093438.1 | - | 8 | 1112 | ||
| AK124125.1 | - | 12 | 1305 | |||
| BC027945.1 | - | 12 | 1326 | |||
| BC126152.1 | - | 8 | 1079 | 8991868 | ||
| BC126154.1 | - | 8 | 1079 | 8991870 | ||

TTLL10
Genomic Map