siRNA Details
| Gene Symbol: TSGA10IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152762.2 | NP_689975.2 | 4 | 1161 | ||
| XM_011544887.2 | XP_011543189.1 | 4 | 1360 | |||
| XM_011544888.2 | XP_011543190.1 | 4 | 1158 | |||
| GenBank mRNA | AK057442.1 | - | 4 | 1161 | ||
| BC104022.1 | - | 4 | 1161 | 40025091 | ||

TSGA10IP
Genomic Map