siRNA Details
| Gene Symbol: TBC1D28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039397.2 | NP_001034486.2 | 10 | 955 | ||
| GenBank mRNA | AK097563.1 | - | 9 | 1243 | ||
| BC112012.1 | - | 8 | 621 | 8327480 | ||
| BC112016.1 | - | 8 | 621 | 8327484 | ||
| Gene Symbol: TBC1D26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK292544.1 | - | 9 | 788 | ||
| AK315847.1 | - | 9 | 788 | |||

TBC1D28
Genomic Map