siRNA Details
| Gene Symbol: PROSER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153256.3 | NP_694988.3 | 3 | 374 | ||
| XM_011519438.2 | XP_011517740.1 | 4 | 693 | |||
| XM_017016028.1 | XP_016871517.1 | 3 | 1360 | |||
| GenBank mRNA | BC017269.2 | - | 3 | 374 | 4588686 | |
| BC048800.1 | - | 3 | 374 | 5215506 | ||

PROSER2
Genomic Map