siRNA Details
| Gene Symbol: SLC41A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173854.5 | NP_776253.3 | 9, 10 | 2249 | ||
| XM_005245069.2 | XP_005245126.1 | 9, 10 | 1925 | |||
| GenBank mRNA | AJ514402.1 | - | 8, 9 | 1336 | ||
| AK093254.1 | - | 1, 2 | 1270 | |||
| AK125075.1 | - | 8, 9 | 1010 | |||
| AK128069.1 | - | 8, 9 | 1502 | |||
| AK314732.1 | - | 8, 9 | 1407 | |||
| AL831974.1 | - | 9, 10 | 2085 | |||
| AL832001.1 | - | 9, 10 | 2085 | |||
| AL832362.1 | - | 9, 10 | 2085 | |||
| BC136303.1 | - | 8, 9 | 1730 | 9020290 | ||
| BX641054.1 | - | 9, 10 | 2088 | |||
| BX648979.1 | - | 9, 10 | 2123 | |||

SLC41A1
Genomic Map