siRNA Details
| Gene Symbol: C8orf46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152765.3 | NP_689978.2 | 1 | 451 | ||
| GenBank mRNA | AK096336.1 | - | 1 | 145 | ||
| AK294261.1 | - | 1 | 101 | |||
| AK308800.1 | - | 1 | 157 | |||
| AK315486.1 | - | 1 | 194 | |||
| BC028400.2 | - | 1 | 175 | 4816822 | ||
| DC344091.1 | - | 1 | 452 | |||

C8orf46
Genomic Map