siRNA Details
| Gene Symbol: C8orf46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152765.3 | NP_689978.2 | 4 | 719 | ||
| GenBank mRNA | AK055047.1 | - | 1 | 53 | ||
| AK096336.1 | - | 5 | 507 | |||
| AK308800.1 | - | 5 | 546 | |||
| AK315486.1 | - | 4 | 462 | |||
| BC028400.2 | - | 4 | 443 | 4816822 | ||

C8orf46
Genomic Map