siRNA Details
| Gene Symbol: OR6C74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005490.1 | NP_001005490.1 | 1 | 63 | ||
| XM_017019126.1 | XP_016874615.1 | 2 | 144 | |||
| GenBank mRNA | BC140754.1 | - | 1 | 153 | 9021622 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR6C74| Gene Symbol: OR6C74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005490.1 | NP_001005490.1 | 1 | 63 | ||
| XM_017019126.1 | XP_016874615.1 | 2 | 144 | |||
| GenBank mRNA | BC140754.1 | - | 1 | 153 | 9021622 | |