siRNA Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207015.2 | NP_996898.2 | 2 | 189 | ||
| XM_006713560.2 | XP_006713623.1 | 4 | 272 | |||
| XM_011512612.2 | XP_011510914.1 | 2 | 3526 | |||
| XM_011512613.1 | XP_011510915.1 | 2 | 334 | |||
| XM_011512615.2 | XP_011510917.1 | 2 | 3527 | |||
| XM_011512617.2 | XP_011510919.1 | 2 | 3524 | |||
| XM_017006070.1 | XP_016861559.1 | 6 | 714 | |||
| XM_017006071.1 | XP_016861560.1 | 7 | 784 | |||
| XM_017006072.1 | XP_016861561.1 | 8 | 918 | |||
| XM_017006073.1 | XP_016861562.1 | 6 | 517 | |||
| XM_017006074.1 | XP_016861563.1 | 8 | 2936 | |||
| XM_017006075.1 | XP_016861564.1 | 5 | 1661 | |||
| XM_017006076.1 | XP_016861565.1 | 5 | 454 | |||
| XM_017006077.1 | XP_016861566.1 | 7 | 1865 | |||
| XM_017006078.1 | XP_016861567.1 | 5 | 1682 | |||
| XM_017006079.1 | XP_016861568.1 | 3 | 1158 | |||
| XM_017006080.1 | XP_016861569.1 | 3 | 1529 | |||
| XM_017006081.1 | XP_016861570.1 | 2 | 3526 | |||
| XM_017006082.1 | XP_016861571.1 | 5 | 1668 | |||
| GenBank mRNA | AJ607396.1 | - | 2 | 230 | ||
| AK308416.1 | - | 2 | 205 | |||
| AL832144.1 | - | 2 | 189 | |||
| BC057243.1 | - | 2 | 192 | |||

NAALADL2
Genomic Map