siRNA Details
| Gene Symbol: LCE5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178438.4 | NP_848525.1 | 2 | 706 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LCE5A| Gene Symbol: LCE5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178438.4 | NP_848525.1 | 2 | 706 | ||