siRNA Details
| Gene Symbol: IGFBP7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034081.1 | 3 | 419 | |||
| GenBank mRNA | BC039519.1 | - | 3 | 419 | ||
| BG189240.1 | - | 2 | 200 | |||
| BG425784.1 | - | 2 | 179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IGFBP7-AS1| Gene Symbol: IGFBP7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034081.1 | 3 | 419 | |||
| GenBank mRNA | BC039519.1 | - | 3 | 419 | ||
| BG189240.1 | - | 2 | 200 | |||
| BG425784.1 | - | 2 | 179 | |||