siRNA Details
| Gene Symbol: MCOLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153259.3 | NP_694991.2 | 13 | 1851 | ||
| XM_005270719.3 | XP_005270776.1 | 13 | 1628 | |||
| XM_006710552.2 | XP_006710615.1 | 14 | 1941 | |||
| XM_011541187.2 | XP_011539489.1 | 13 | 1685 | |||
| XM_011541188.2 | XP_011539490.1 | 13 | 1968 | |||
| GenBank mRNA | AK094010.1 | - | 11 | 1526 | ||
| AK129786.1 | - | 7 | 854 | |||
| AY083533.1 | - | 12 | 1517 | |||
| BC104891.1 | - | 13 | 1669 | 8143894 | ||
| BC104893.1 | - | 13 | 1669 | 8143896 | ||

MCOLN2
Genomic Map