siRNA Details
| Gene Symbol: ANKK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178510.1 | NP_848605.1 | 2 | 344 | ||
| XM_011542736.2 | XP_011541038.1 | 2 | 2121 | |||
| XM_011542737.2 | XP_011541039.1 | 2 | 2121 | |||
| XM_011542738.2 | XP_011541040.1 | 2 | 2379 | |||
| XM_017017475.1 | XP_016872964.1 | 2 | 2121 | |||
| GenBank mRNA | AF487542.1 | - | 2 | 250 | ||
| AJ541797.2 | - | 2 | 344 | |||

ANKK1
Genomic Map