siRNA Details
| Gene Symbol: EPGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270989.1 | NP_001257918.1 | 4 | 360 | ||
| XM_005265666.4 | XP_005265723.1 | 4 | 1232 | |||
| XM_011531817.2 | XP_011530119.1 | 4 | 1218 | |||
| GenBank mRNA | AK289455.1 | - | 4 | 358 | ||
| AY358920.1 | - | 4 | 316 | |||
| CB150835.1 | - | 4 | 333 | |||
| DQ235269.1 | - | 4 | 344 | |||

EPGN
Genomic Map