siRNA Details
| Gene Symbol: MBLAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203397.2 | NP_981942.1 | 2 | 1445 | ||
| XM_005250250.3 | XP_005250307.1 | 2 | 1488 | |||
| GenBank mRNA | BC031288.1 | - | 2 | 1442 | 5299411 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MBLAC1| Gene Symbol: MBLAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203397.2 | NP_981942.1 | 2 | 1445 | ||
| XM_005250250.3 | XP_005250307.1 | 2 | 1488 | |||
| GenBank mRNA | BC031288.1 | - | 2 | 1442 | 5299411 | |