siRNA Details
| Gene Symbol: RASGEF1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175062.3 | NP_778232.2 | 11 | 1275 | ||
| GenBank mRNA | AF449764.1 | - | 10 | 1463 | ||
| AK093160.1 | - | 8 | 1127 | |||
| BC036802.1 | - | 12 | 1293 | |||
| BC057759.1 | - | 9 | 967 | 5268021 | ||
| BM729117.1 | - | 2 | 116 | |||

RASGEF1C
Genomic Map