siRNA Details
| Gene Symbol: EVC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306090.1 | NP_001293019.1 | 6 | 961 | ||
| NM_001306092.1 | NP_001293021.1 | 6 | 961 | |||
| NM_153717.2 | NP_714928.1 | 6 | 961 | |||
| XM_006713865.3 | XP_006713928.1 | 6 | 958 | |||
| XM_006713866.3 | XP_006713929.1 | 6 | 958 | |||
| XM_011513419.2 | XP_011511721.1 | 6 | 955 | |||
| XM_017007883.1 | XP_016863372.1 | 6 | 955 | |||

EVC
Genomic Map