siRNA Details
| Gene Symbol: ANKRD31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164443.1 | NP_001157915.1 | 4, 5 | 516 | ||
| XM_011543298.2 | XP_011541600.1 | 4, 5 | 538 | |||
| XM_011543299.1 | XP_011541601.1 | 4, 5 | 516 | |||
| XM_011543300.2 | XP_011541602.1 | 4, 5 | 538 | |||
| XM_011543301.2 | XP_011541603.1 | 4, 5 | 538 | |||
| XM_011543302.2 | XP_011541604.1 | 2, 3 | 74 | |||
| XM_017009317.1 | XP_016864806.1 | 4, 5 | 433 | |||
| XM_017009318.1 | XP_016864807.1 | 4, 5 | 538 | |||
| XM_017009319.1 | XP_016864808.1 | 4, 5 | 538 | |||
| XM_017009322.1 | XP_016864811.1 | 4, 5 | 538 | |||
| GenBank mRNA | AK097510.1 | - | 4, 5 | 516 | ||

ANKRD31
Genomic Map