siRNA Details
Gene Symbol: ZNF549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199295.1 | NP_001186224.1 | 4 | 1783 | ||
NM_153263.2 | NP_694995.2 | 3 | 1744 | |||
XM_006723128.3 | XP_006723191.1 | 4 | 1858 | |||
GenBank mRNA | AK092236.1 | - | 3 | 1744 | ||
AK094139.1 | - | 1 | 238 | |||
AK122749.1 | - | 4 | 1717 | |||
AK122766.1 | - | 1 | 1034 | |||
BC060863.1 | - | 4 | 1687 | 30340439 |
