siRNA Details
| Gene Symbol: GCSAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190259.1 | NP_001177188.1 | 4 | 371 | ||
| NM_001190260.1 | NP_001177189.1 | 3 | 320 | |||
| NM_152785.4 | NP_689998.1 | 4 | 365 | |||
| XM_005247272.2 | XP_005247329.1 | 4 | 789 | |||
| XM_011512627.2 | XP_011510929.1 | 4 | 240 | |||
| XM_011512628.2 | XP_011510930.1 | 4 | 297 | |||
| XM_011512629.1 | XP_011510931.1 | 4 | 789 | |||
| XM_011512630.1 | XP_011510932.1 | 4 | 795 | |||
| GenBank mRNA | AF521911.1 | - | 4 | 352 | ||
| AK313368.1 | - | 4 | 312 | |||
| AY212246.1 | - | 4 | 286 | |||
| BC030506.1 | - | 4 | 233 | 4385178 | ||
| BM456595.1 | - | 4 | 321 | |||
| BP224611.1 | - | 3 | 268 | |||
| BX280641.1 | - | 4 | 421 | |||
| DA455714.1 | - | 4 | 366 | |||

GCSAM
Genomic Map