siRNA Details
| Gene Symbol: GCSAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190259.1 | NP_001177188.1 | 3 | 304 | ||
| NM_152785.4 | NP_689998.1 | 3 | 298 | |||
| XM_005247272.2 | XP_005247329.1 | 3 | 722 | |||
| XM_011512627.2 | XP_011510929.1 | 3 | 173 | |||
| XM_011512628.2 | XP_011510930.1 | 3 | 230 | |||
| XM_011512629.1 | XP_011510931.1 | 3 | 722 | |||
| XM_011512630.1 | XP_011510932.1 | 3 | 728 | |||
| GenBank mRNA | AF521911.1 | - | 3 | 285 | ||
| AK313368.1 | - | 3 | 245 | |||
| AY212246.1 | - | 3 | 219 | |||
| BC030506.1 | - | 3 | 166 | 4385178 | ||
| BM456595.1 | - | 3 | 254 | |||
| BX280641.1 | - | 3 | 354 | |||
| DA455714.1 | - | 3 | 299 | |||

GCSAM
Genomic Map