siRNA Details
| Gene Symbol: RNF214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077239.1 | NP_001070707.1 | 14, 15 | 2091 | ||
| NM_001278249.1 | NP_001265178.1 | 14, 15 | 1626 | |||
| NM_207343.3 | NP_997226.2 | 14, 15 | 2153 | |||
| GenBank mRNA | AK298923.1 | - | 14, 15 | 2085 | ||
| AK300160.1 | - | 14, 15 | 1626 | |||
| AL834448.1 | - | 4, 5 | 633 | |||
| BC031347.1 | - | 9, 10 | 1188 | |||
| BC146891.1 | - | 14, 15 | 2091 | 9056819 | ||
| BC146901.1 | - | 14, 15 | 2091 | 9056829 | ||
| BX111206.1 | - | 2, 3 | 124 | |||

RNF214
Genomic Map