siRNA Details
| Gene Symbol: C2orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144994.1 | NP_001138466.1 | 3 | 1819 | ||
| GenBank mRNA | AF007149.1 | - | 2 | 1306 | ||
| AF070612.1 | - | 1 | 349 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf72| Gene Symbol: C2orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144994.1 | NP_001138466.1 | 3 | 1819 | ||
| GenBank mRNA | AF007149.1 | - | 2 | 1306 | ||
| AF070612.1 | - | 1 | 349 | |||