siRNA Details
| Gene Symbol: TMIE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147196.2 | NP_671729.2 | 4 | 1391 | ||
| XM_006713097.3 | XP_006713160.1 | 4 | 1962 | |||
| GenBank mRNA | AY081842.1 | - | 4 | 1388 | ||
| BM696323.1 | - | 1 | 160 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMIE| Gene Symbol: TMIE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147196.2 | NP_671729.2 | 4 | 1391 | ||
| XM_006713097.3 | XP_006713160.1 | 4 | 1962 | |||
| GenBank mRNA | AY081842.1 | - | 4 | 1388 | ||
| BM696323.1 | - | 1 | 160 | |||