siRNA Details
| Gene Symbol: WFDC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147197.2 | NP_671730.1 | 3, 4 | 289 | ||
| GenBank mRNA | AY047609.1 | - | 3, 4 | 289 | ||
| BC062670.1 | - | 3, 4 | 320 | 6101449 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WFDC11| Gene Symbol: WFDC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147197.2 | NP_671730.1 | 3, 4 | 289 | ||
| GenBank mRNA | AY047609.1 | - | 3, 4 | 289 | ||
| BC062670.1 | - | 3, 4 | 320 | 6101449 | ||