siRNA Details
| Gene Symbol: TAS2R39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176881.2 | NP_795362.2 | 1 | 301 | ||
| GenBank mRNA | BC100947.2 | - | 1 | 301 | ||
| BC100948.2 | - | 1 | 301 | |||
| BC114952.1 | - | 1 | 301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R39| Gene Symbol: TAS2R39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176881.2 | NP_795362.2 | 1 | 301 | ||
| GenBank mRNA | BC100947.2 | - | 1 | 301 | ||
| BC100948.2 | - | 1 | 301 | |||
| BC114952.1 | - | 1 | 301 | |||