siRNA Details
Gene Symbol: TAS2R40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176882.1 | NP_795363.1 | 1 | 373 | ||
GenBank mRNA | BC101795.1 | - | 1 | 415 | 8069301 | |
BC101797.1 | - | 1 | 415 | 8069303 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TAS2R40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176882.1 | NP_795363.1 | 1 | 373 | ||
GenBank mRNA | BC101795.1 | - | 1 | 415 | 8069301 | |
BC101797.1 | - | 1 | 415 | 8069303 |