siRNA Details
Gene Symbol: TAS2R43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176884.2 | NP_795365.2 | 1 | 592 | ||
GenBank mRNA | BC117423.1 | - | 1 | 591 | 40125974 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TAS2R43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176884.2 | NP_795365.2 | 1 | 592 | ||
GenBank mRNA | BC117423.1 | - | 1 | 591 | 40125974 |