siRNA Details
Gene Symbol: TAS2R45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176886.2 | NP_795367.2 | 2 | 183 | ||
GenBank mRNA | BC101154.1 | - | 2 | 183 | 40015678 | |
BC101156.1 | - | 2 | 183 | 40015680 |
Gene Symbol: TAS2R43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC101155.1 | - | 2 | 183 | 40015679 |
