siRNA Details
| Gene Symbol: TAS2R46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176887.2 | NP_795368.2 | 1 | 55 | ||
| GenBank mRNA | BC115401.2 | - | 1 | 55 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R46| Gene Symbol: TAS2R46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176887.2 | NP_795368.2 | 1 | 55 | ||
| GenBank mRNA | BC115401.2 | - | 1 | 55 | ||