siRNA Details
| Gene Symbol: TAS2R20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176889.2 | NP_795370.2 | 1 | 673 | ||
| GenBank mRNA | BC100915.2 | - | 1 | 673 | 40003843 | |
| BC100916.2 | - | 1 | 673 | 40003844 | ||
| BC100917.2 | - | 1 | 673 | 40003847 | ||
| BC100918.2 | - | 1 | 673 | 40003851 | ||
| BC113843.1 | - | 1 | 673 | 40003846 | ||
| BC114451.1 | - | 1 | 673 | 40003845 | ||

TAS2R20
Genomic Map