siRNA Details
| Gene Symbol: HS6ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153456.3 | NP_703157.2 | 2 | 1248 | ||
| GenBank mRNA | AF539426.1 | - | 2 | 1248 | ||
| AK126947.1 | - | 2 | 750 | |||
| CR749677.1 | - | 2 | 914 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS6ST3| Gene Symbol: HS6ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153456.3 | NP_703157.2 | 2 | 1248 | ||
| GenBank mRNA | AF539426.1 | - | 2 | 1248 | ||
| AK126947.1 | - | 2 | 750 | |||
| CR749677.1 | - | 2 | 914 | |||