siRNA Details
| Gene Symbol: MDGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153487.3 | NP_705691.1 | 6 | 1913 | ||
| XM_006715056.3 | XP_006715119.1 | 6 | 857 | |||
| XM_017010734.1 | XP_016866223.1 | 6 | 857 | |||
| GenBank mRNA | AF478693.1 | - | 6 | 1913 | ||
| AK091149.1 | - | 4 | 1317 | |||

MDGA1
Genomic Map