siRNA Details
| Gene Symbol: WFDC10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172006.2 | NP_742003.1 | 4 | 358 | ||
| NM_172131.2 | NP_742143.1 | 3 | 203 | |||
| XM_017027824.1 | XP_016883313.1 | 5 | 430 | |||
| XM_017027825.1 | XP_016883314.1 | 5 | 683 | |||
| GenBank mRNA | AF454506.1 | - | 4 | 358 | ||
| AF454507.1 | - | 3 | 203 | |||
| BC121071.1 | - | 4 | 356 | 40119593 | ||
| BC121072.1 | - | 4 | 356 | 40119597 | ||

WFDC10B
Genomic Map