siRNA Details
| Gene Symbol: AQP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173039.2 | NP_766627.1 | 2 | 1024 | ||
| GenBank mRNA | AB028147.1 | - | 2 | 971 | ||
| AK075346.1 | - | 2 | 666 | |||
| AK311096.1 | - | 3 | 421 | |||
| BC040443.1 | - | 2 | 1024 | 5728157 | ||

AQP11
Genomic Map