siRNA Details
| Gene Symbol: POC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199777.1 | NP_001186706.1 | 10 | 1634 | ||
| NM_172240.2 | NP_758440.1 | 11 | 1653 | |||
| NR_037659.1 | 11 | 1504 | ||||
| NR_037660.1 | 9 | 1462 | ||||
| GenBank mRNA | AB053266.1 | - | 10 | 1143 | ||
| AK027829.1 | - | 7 | 852 | |||
| AK074772.1 | - | 11 | 1391 | |||
| AK123106.1 | - | 10 | 1631 | |||
| AK123625.1 | - | 7 | 851 | |||
| AK295596.1 | - | 11 | 1242 | |||
| AL832918.1 | - | 10 | 1140 | |||
| BC026080.2 | - | 11 | 1415 | 4827689 | ||
| BX648293.1 | - | 1 | 4255 | |||
| EF011619.1 | - | 11 | 1415 | |||

POC1B
Genomic Map