siRNA Details
| Gene Symbol: EYA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005244.4 | NP_005235.3 | 5, 6 | 798 | ||
| NM_172110.3 | NP_742108.2 | 5, 6 | 798 | |||
| XM_017027721.1 | XP_016883210.1 | 5, 6 | 516 | |||
| GenBank mRNA | AF055015.1 | - | 1, 2 | 112 | ||
| AK293032.1 | - | 5, 6 | 502 | |||
| AY705349.1 | - | 4, 5 | 434 | |||
| BC008803.2 | - | 5, 6 | 480 | 3944547 | ||
| BC013882.2 | - | 5, 6 | 798 | |||
| BT006682.1 | - | 4, 5 | 352 | |||
| U71207.1 | - | 4, 5 | 426 | |||

EYA2
Genomic Map