siRNA Details
| Gene Symbol: C10orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042427.1 | NP_001035892.1 | 2 | 229 | ||
| NM_182554.2 | NP_872360.2 | 2 | 229 | |||
| GenBank mRNA | AK091773.1 | - | 2 | 216 | ||
| BC028127.1 | - | 2 | 181 | 5164374 | ||
| BX102324.1 | - | 2 | 229 | |||
| DB043416.1 | - | 2 | 202 | |||

C10orf53
Genomic Map