siRNA Details
| Gene Symbol: LINC00900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034148.1 | 2 | 270 | |||
| GenBank mRNA | AK090602.1 | - | 2 | 264 | ||
| AK307308.1 | - | 2 | 304 | |||
| AK309234.1 | - | 3 | 279 | |||
| DA991791.1 | - | 2 | 270 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00900| Gene Symbol: LINC00900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034148.1 | 2 | 270 | |||
| GenBank mRNA | AK090602.1 | - | 2 | 264 | ||
| AK307308.1 | - | 2 | 304 | |||
| AK309234.1 | - | 3 | 279 | |||
| DA991791.1 | - | 2 | 270 | |||