siRNA Details
| Gene Symbol: BCL9L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182557.2 | NP_872363.1 | 6 | 2548 | ||
| XM_005271511.2 | XP_005271568.1 | 8 | 2548 | |||
| XM_005271512.2 | XP_005271569.1 | 7 | 1998 | |||
| XM_005271513.2 | XP_005271570.1 | 6 | 1997 | |||
| XM_006718815.2 | XP_006718878.1 | 7 | 2046 | |||
| XM_011542756.2 | XP_011541058.1 | 6 | 2504 | |||
| XM_017017572.1 | XP_016873061.1 | 8 | 2797 | |||
| XM_017017573.1 | XP_016873062.1 | 7 | 3149 | |||
| XM_017017574.1 | XP_016873063.1 | 7 | 2696 | |||
| GenBank mRNA | AB094091.1 | - | 6 | 2548 | ||
| AK122882.1 | - | 1 | 188 | |||
| AY296059.1 | - | 6 | 1568 | |||

BCL9L
Genomic Map