siRNA Details
| Gene Symbol: FOXR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181721.2 | NP_859072.1 | 3 | 394 | ||
| XM_017017575.1 | XP_016873064.1 | 3 | 699 | |||
| XM_017017576.1 | XP_016873065.1 | 3 | 703 | |||
| GenBank mRNA | AB094092.1 | - | 3 | 394 | ||
| BC028191.1 | - | 3 | 393 | 5167039 | ||
| BC038969.1 | - | 3 | 269 | 5164198 | ||
| BC125040.1 | - | 3 | 389 | 40115641 | ||

FOXR1
Genomic Map