siRNA Details
| Gene Symbol: OR8D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002918.1 | NP_001002918.1 | 1 | 860 | ||
| GenBank mRNA | AF162668.1 | - | 1 | 860 | ||
| BC137043.1 | - | 1 | 951 | 9021039 | ||
| BC137045.1 | - | 1 | 951 | 9021041 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR8D2| Gene Symbol: OR8D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002918.1 | NP_001002918.1 | 1 | 860 | ||
| GenBank mRNA | AF162668.1 | - | 1 | 860 | ||
| BC137043.1 | - | 1 | 951 | 9021039 | ||
| BC137045.1 | - | 1 | 951 | 9021041 | ||