siRNA Details
| Gene Symbol: OR8B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005196.1 | NP_001005196.1 | 1 | 581 | ||
| GenBank mRNA | BC136865.1 | - | 1 | 603 | 9020856 | |
| BC136866.1 | - | 1 | 603 | 9020857 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR8B4| Gene Symbol: OR8B4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005196.1 | NP_001005196.1 | 1 | 581 | ||
| GenBank mRNA | BC136865.1 | - | 1 | 603 | 9020856 | |
| BC136866.1 | - | 1 | 603 | 9020857 | ||