siRNA Details
| Gene Symbol: HARBI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173811.3 | NP_776172.1 | 3 | 932 | ||
| XM_011520025.1 | XP_011518327.1 | 4 | 1016 | |||
| XM_011520026.2 | XP_011518328.1 | 4 | 993 | |||
| XM_011520027.1 | XP_011518329.1 | 4 | 1027 | |||
| XM_011520028.2 | XP_011518330.1 | 4 | 1002 | |||
| XM_011520029.2 | XP_011518331.1 | 4 | 1288 | |||
| XM_017017602.1 | XP_016873091.1 | 4 | 1132 | |||
| XM_017017603.1 | XP_016873092.1 | 4 | 1047 | |||
| XM_017017604.1 | XP_016873093.1 | 3 | 943 | |||
| XM_017017605.1 | XP_016873094.1 | 3 | 1204 | |||
| XM_017017606.1 | XP_016873095.1 | 3 | 959 | |||
| GenBank mRNA | AK057237.1 | - | 3 | 932 | ||
| BC036925.1 | - | 3 | 864 | 5225925 | ||

HARBI1
Genomic Map